Dimethyl phenylphosphonite 98%

Supplier: Ambeed
Danger

Synonyms: Phenyldimethoxyphosphine

A284819-1G A284819-5G A284819-25G
76802-278EA 7 USD
76802-278 76802-280 77550-750
Dimethyl phenylphosphonite 98%
Dimethyl phenylphosphonite

Precaución: for Research Use Only

Formula: C₈H₁₁O₂P
MDL Number: MFCD00008352
CAS Number: 2946-61-4
UN: 1760
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR