Di-µ-chlorodichlorobis(ethylene)diplatinum(II) 98%

Supplier: Ambeed
Danger

A284454-250MG A284454-1G A284454-5G A284454-100MG
76802-272EA 108 USD
76802-272 77626-456 77626-458 77879-551
Di-µ-chlorodichlorobis(ethylene)diplatinum(II) 98%
Di-µ-chlorodichlorobis(ethylene)diplatinum(II)

Precaución: for Research Use Only

Formula: C₄H₄Cl₄Pt₂
MDL Number: MFCD00058713
CAS Number: 12073-36-8
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR