1-Methyl-3-phenylpiperazine 98%

Supplier: Ambeed
Danger

Synonyms: MEFPIP

A283272-100G A283272-10G A283272-1G A283272-25G A283272-500G A283272-5G
76802-246EA 120.95 USD
76802-246 76802-248 76802-250 76802-252 76802-254 76802-256
1-Methyl-3-phenylpiperazine 98%
1-Methyl-3-phenylpiperazine

Precaución: for Research Use Only

Formula: C₁₁H₁₆N₂
MW: 176.26 g/mol
MDL Number: MFCD03411603
CAS Number: 5271-27-2
UN: 2923
ADR: 8,6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR