3-Phenylaniline 98%

Supplier: Ambeed
Warning

A283172-100G A283172-10G A283172-1G A283172-25G A283172-500G A283172-5G
76802-234EA 83 USD
76802-234 76802-236 76802-238 76802-240 76802-242 76802-244
3-Phenylaniline 98%
3-Phenylaniline

Precaución: for Research Use Only

Formula: C₁₂H₁₁N
MDL Number: MFCD00047846
CAS Number: 2243-47-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR