(Trimethoxysilyl)methyl methacrylate 97%

Supplier: Ambeed
Warning

A282988-25G A282988-100G A282988-1G A282988-5G
76802-232EA 201 USD
76802-232 77879-541 77879-542 77879-543
(Trimethoxysilyl)methyl methacrylate 97%
(Trimethoxysilyl)methyl methacrylate

Precaución: for Research Use Only

Formula: C₈H₁₆O₅Si
MDL Number: MFCD00156471
CAS Number: 54586-78-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR