(S)-2-Amino-2-(thiophen-3-yl)acetic acid 97%

Supplier: Ambeed
Warning

Synonyms: H-α-(3-Thienyl)-Gly-OH

A282645-1G A282645-250MG
76802-228EA 399.96 USD
76802-228 76802-230
(S)-2-Amino-2-(thiophen-3-yl)acetic acid 97%
(S)-2-Amino-2-(thiophen-3-yl)acetic acid

Precaución: for Research Use Only

Formula: C₆H₇NO₂S
MW: 157.19 g/mol
MDL Number: MFCD00079617
CAS Number: 1194-87-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR