DL-Valine ethyl ester hydrochloride 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
H-DL-Val-OEt·HCl, H-DL-Val-OEt.HCl
A282306-100G
A282306-10G
A282306-1G
A282306-25G
A282306-5G
76802-214EA
103
USD
76802-214
76802-216
76802-218
76802-220
76802-222
DL-Valine ethyl ester hydrochloride 98%
DL-Valine ethyl ester hydrochloride
Precaución: for Research Use Only
Formula:
C₇H₁₅NO₂·HCl |
MDL Number:
MFCD00067542 CAS Number: 23358-42-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...