3,5-Bis(benzyloxy)benzaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 3,5-Dibenzyloxybenzaldehyde

A281368-1G A281368-5G
76802-200EA 9.55 USD
76802-200 77593-738
3,5-Bis(benzyloxy)benzaldehyde 98%
3,5-Bis(benzyloxy)benzaldehyde

Precaución: for Research Use Only

Formula: C₂₁H₁₈O₃
MW: 318.3658 g/mol
MDL Number: MFCD00075777
CAS Number: 14615-72-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR