D-(-)-Lactic acid 90% (by titrimetric analysis)
Supplier: Ambeed
|
Warning
|
A278821-25G
A278821-100G
A278821-500G
76802-158EA
46.68
USD
76802-158
76802-160
77568-550
77601-230
D-(-)-Lactic acid 90% (by titrimetric analysis)
D-(-)-Lactic acid
Precaución: For research use only.
Formula:
C₃H₆O₃ Storage Temperature: Freezer |
MDL Number:
MFCD00068313 CAS Number: 10326-41-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...