Dipropyleneglycol monomethyl ether (mixture of isomers) 98% (mixture of isomers)

Supplier: Ambeed
Warning

A277119-100G A277119-500G
76802-126EA 8 USD
76802-126 77879-516
Dipropyleneglycol monomethyl ether (mixture of isomers) 98% (mixture of isomers)
Dipropyleneglycol monomethyl ether (mixture of isomers)

Precaución: for Research Use Only

Formula: C₇H₁₆O₃
MDL Number: MFCD00059604
CAS Number: 34590-94-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR