Ethyl-2-oxohexanoate 96%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-oxohexanoate

A276711-10G A276711-1G A276711-25G A276711-5G
76802-120EA 39 USD
76802-120 76802-122 76802-124 77879-513
Ethyl-2-oxohexanoate 96%
Ethyl-2-oxohexanoate

Precaución: for Research Use Only

Formula: C₈H₁₄O₃
MDL Number: MFCD09952021
CAS Number: 5753-96-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR