Diamminedichloropalladium(II) 98%

Supplier: Ambeed
Danger

A275781-250MG A275781-100MG
76802-106EA 53 USD
76802-106 77879-512
Diamminedichloropalladium(II) 98%
Diamminedichloropalladium(II)

Precaución: for Research Use Only

Formula: Cl₂H₆N₂Pd
MDL Number: MFCD00011621
CAS Number: 14323-43-4
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR