Febuxostat 98%

Supplier: Ambeed
Warning

Synonyms: 2-(3-Cyano-4-isobutoxyphenyl)-4-methylthiazole-5-carboxylic acid

A274351-100G A274351-10G A274351-1G A274351-25G A274351-50MG A274351-5G A274351-100MG A274351-500G
76802-060EA 192.03 USD
76802-060 76802-062 76802-064 76802-066 76802-068 76802-070 77593-694 77668-200
Febuxostat 98%
Febuxostat

Precaución: for Research Use Only

Formula: C₁₆H₁₆N₂O₃S
MW: 316.37 g/mol
MDL Number: MFCD00871598
CAS Number: 144060-53-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR