H-DL-Glu(Ome)-OH 98%

Supplier: Ambeed
Warning

A274316-25G A274316-5G
76802-056EA 58.35 USD
76802-056 76802-058
H-DL-Glu(Ome)-OH 98%
H-DL-Glu(Ome)-OH

Precaución: for Research Use Only

Formula: C₆H₁₁NO₄
MW: 161.15 g/mol
MDL Number: MFCD00063110
CAS Number: 14487-45-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR