Ethyl 2-(3-fluoro-4-nitrophenyl)propanoate 97%

Supplier: Ambeed
Warning

A273657-100MG A273657-1G A273657-250MG A273657-5G
76802-020EA 45 USD
76802-020 76802-022 76802-024 76802-026
Ethyl 2-(3-fluoro-4-nitrophenyl)propanoate 97%
Ethyl 2-(3-fluoro-4-nitrophenyl)propanoate

Precaución: for Research Use Only

Formula: C₁₁H₁₂FNO₄
MDL Number: MFCD11053617
CAS Number: 78543-07-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR