Methacroylcholine chloride in water ≥80% stabilised

Supplier: Ambeed
Warning

Synonyms: 2-(Methacryloyloxy)-N,N,N-trimethylethanaminium chloride, (Methacryloyloxyethyl)trimethyl ammonium chloride, 2-(Methacryloyloxy)ethyltrimethylammonium chloride, Trimethyl-2-methacroyloxyethylammonium Chloride

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A272811-100G A272811-500G A272811-25G A272811-1000G
76802-012EA 9 USD
76802-012 76802-014 77092-810 77879-504
Methacroylcholine chloride in water ≥80% stabilised
Methacroylcholine chloride

Precaución: for Research Use Only

Formula: C₉H₁₈ClNO₂
MDL Number: MFCD00060097
CAS Number: 5039-78-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR