5-Amino-1-(4-methylphenyl)-1H-pyrazole-4-carbonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 5-Amino-1-(p-tolyl)-1H-pyrazole-4-carbonitrile, 5-Amino-1-(4-methylphenyl)pyrazole-4-carbonitrile, 5-Amino-1-(p-tolyl)pyrazole-4-carbonitrile

A270951-1G A270951-250MG A270951-5G
76801-982EA 51.99 USD
76801-982 76801-984 76801-986
5-Amino-1-(4-methylphenyl)-1H-pyrazole-4-carbonitrile 97%
5-Amino-1-(4-methylphenyl)-1H-pyrazole-4-carbonitrile

Precaución: for Research Use Only

Formula: C₁₁H₁₀N₄
MW: 198.23 g/mol
MDL Number: MFCD00052944
CAS Number: 103646-82-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR