4-(4-Fluorobenzoyl)butyric acid 98%

Supplier: Ambeed
Warning

Synonyms: 5-(4-Fluorophenyl)-5-oxovaleric Acid, 5-(4-Fluorophenyl)-5-oxopentanoic acid

A269940-1000G A269940-100G A269940-10G A269940-25G A269940-500G
76801-958EA 230.22 USD
76801-958 76801-960 76801-962 76801-964 76801-966
4-(4-Fluorobenzoyl)butyric acid 98%
4-(4-Fluorobenzoyl)butyric acid

Precaución: for Research Use Only

Formula: C₁₁H₁₁FO₃
MW: 210.20 g/mol
MDL Number: MFCD00667222
CAS Number: 149437-76-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR