manidipine 98%

Supplier: Ambeed
Warning

Synonyms: 3-(2-(4-Benzhydrylpiperazin-1-yl)ethyl) 5-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate

A268936-1G A268936-250MG A268936-5G
76801-948EA 33.95 USD
76801-948 76801-950 76801-952
manidipine 98%
manidipine

Precaución: for Research Use Only

Formula: C₃₅H₃₈N₄O₆
MW: 610.711 g/mol
MDL Number: MFCD00871291
CAS Number: 89226-50-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR