6-Methylheptyl methacrylate 99%

Supplier: Ambeed
Warning

A268852-100G A268852-25G A268852-500G
76801-942EA 61 USD
76801-942 76801-944 76801-946
6-Methylheptyl methacrylate 99%
6-Methylheptyl methacrylate

Precaución: for Research Use Only

Formula: C₁₂H₂₂O₂
MW: 198.30 g/mol
CAS Number: 28675-80-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR