Methyl-1,5-dimethyl-1H-pyrazole-3-carboxylate 98+%

Supplier: Ambeed
Warning

A267692-5G A267692-1G A267692-250MG
76801-934EA 150 USD
76801-934 77879-495 77879-496
Methyl-1,5-dimethyl-1H-pyrazole-3-carboxylate 98+%
Methyl-1,5-dimethyl-1H-pyrazole-3-carboxylate

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O₂
MDL Number: MFCD06661312
CAS Number: 10250-61-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR