Sodium meclofenamate hydrate 99%

Supplier: Ambeed
Warning

Synonyms: Meclofenamic acid Sodium salt, 2-[(2,6-Dichloro-3-methylphenyl)amino]benzoic acid Sodium salt, Sodium 2-((2,6-dichloro-3-methylphenyl)amino)benzoate

A267516-10MG A267516-1G A267516-1MG A267516-250MG A267516-50MG A267516-100MG
76801-920EA 7.43 USD
76801-920 76801-922 76801-924 76801-926 76801-928 77668-158
Sodium meclofenamate hydrate 99%
Sodium meclofenamate hydrate

Precaución: for Research Use Only

Formula: C₁₄H₁₁Cl₂NO₂
MW: 318.13 g/mol
MDL Number: MFCD00077376
CAS Number: 6385-02-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR