Triethyl-1,1,2-ethanetricarboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Triethyl ethane-1,1,2-tricarboxylate

A265908-100G A265908-25G A265908-500G
76801-908EA 115 USD
76801-908 76801-910 76801-912
Triethyl-1,1,2-ethanetricarboxylate 98%
Triethyl-1,1,2-ethanetricarboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₈O₆
MW: 246.26 g/mol
MDL Number: MFCD00009154
CAS Number: 7459-46-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR