(S)-6-Amino-2-((S)-2-(2-aminoacetamido)-3-(1H-imidazol-4-yl)propanamido)hexanoic acid 98%

Supplier: Ambeed
Warning

Synonyms: Glycyl-L-histidyl-L-lysine

A265833-1G A265833-1MG A265833-250MG A265833-5G
76801-900EA 39.26 USD
76801-900 76801-902 76801-904 76801-906
(S)-6-Amino-2-((S)-2-(2-aminoacetamido)-3-(1H-imidazol-4-yl)propanamido)hexanoic acid 98%
(S)-6-Amino-2-((S)-2-(2-aminoacetamido)-3-(1H-imidazol-4-yl)propanamido)hexanoic acid

Precaución: for Research Use Only

Formula: C₁₄H₂₄N₆O₄
MW: 340.38 g/mol
MDL Number: MFCD00036754
CAS Number: 49557-75-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR