Furfuryl acetate 98%

Supplier: Ambeed
Warning

Synonyms: Furan-2-ylmethyl acetate, Acetic acid furfuryl ester

A265513-100G A265513-10G A265513-500G A265513-25G
76801-874EA 23.34 USD
76801-874 76801-876 76801-878 77092-322
Furfuryl acetate 98%
Furfuryl acetate

Precaución: for Research Use Only

Formula: C₇H₈O₃
MW: 140.14 g/mol
MDL Number: MFCD00003251
CAS Number: 623-17-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR