Pentafluoroiodobenzene 98% stabilized

Supplier: Ambeed
Warning

Synonyms: Iodopentafluorobenzene

Stabilization: Stabilized with Copper chip

A265105-10G A265105-1G A265105-25G A265105-5G A265105-100G
76801-860EA 36.08 USD
76801-860 76801-862 76801-864 76801-866 77550-664
Pentafluoroiodobenzene 98% stabilized
Pentafluoroiodobenzene

Precaución: for Research Use Only

Formula: C₆F₅I
MW: 293.96 g/mol
MDL Number: MFCD00001032
CAS Number: 827-15-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR