Diethyl 2,6-Dimethylpyridine-3,5-dicarboxylate 97%

Supplier: Ambeed
Warning

A265064-100MG A265064-10G A265064-1G A265064-250MG A265064-5G
76801-850EA 7 USD
76801-850 76801-852 76801-854 76801-856 76801-858
Diethyl 2,6-Dimethylpyridine-3,5-dicarboxylate 97%
Diethyl 2,6-Dimethylpyridine-3,5-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₃H₁₇NO₄
MDL Number: MFCD00023506
CAS Number: 1149-24-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR