N-Fmoc-8-aminooctanoic acid 97%

Supplier: Ambeed
Warning

Synonyms: Fmoc-8-Aoc-OH, Fmoc-8-aminooctanoic acid

A264170-1G A264170-250MG A264170-25G A264170-5G
76801-838EA 18 USD
76801-838 76801-840 76801-842 77550-660
N-Fmoc-8-aminooctanoic acid 97%
N-Fmoc-8-aminooctanoic acid

Precaución: for Research Use Only

Formula: C₂₃H₂₇NO₄
MDL Number: MFCD00235890
CAS Number: 126631-93-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR