Ethyl 2-(4-hydroxypyrimidin-5-yl)acetate 96%

Supplier: Ambeed
Warning

A263932-100MG A263932-10G A263932-250MG A263932-1G A263932-5G
76801-832EA 30.77 USD
76801-832 76801-834 76801-836 77668-134 77668-136
Ethyl 2-(4-hydroxypyrimidin-5-yl)acetate 96%
Ethyl 2-(4-hydroxypyrimidin-5-yl)acetate

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O₃
MW: 182.177 g/mol
MDL Number: MFCD11656636
CAS Number: 6214-46-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR