Norharmane 98%

Supplier: Ambeed
Warning

Synonyms: Norharman, 9H-Pyrido[3,4-b]indole

A262431-10G A262431-1G A262431-250MG A262431-25G A262431-5G
76801-810EA 343.74 USD
76801-810 76801-812 76801-814 76801-816 76801-818
Norharmane 98%
Norharmane

Precaución: for Research Use Only

Formula: C₁₁H₈N₂
MW: 168.2 g/mol
MDL Number: MFCD00004956
CAS Number: 244-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR