1-Bromo-2-naphthoic acid 97%

Supplier: Ambeed
Warning

Synonyms: 1-Bromonaphthalene-2-carboxylic acid

A262360-100G A262360-10G A262360-1G A262360-250MG A262360-25G A262360-5G
76801-798EA 595.17 USD
76801-798 76801-800 76801-802 76801-804 76801-806 76801-808
1-Bromo-2-naphthoic acid 97%
1-Bromo-2-naphthoic acid

Precaución: for Research Use Only

Formula: C₁₁H₇BrO₂
MW: 251.0800 g/mol
MDL Number: MFCD00021408
CAS Number: 20717-79-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR