Isopropyl myristate 98%

Supplier: Ambeed
Warning

A260428-100G A260428-500G A260428-1000G
76801-778EA 13 USD
76801-778 76801-780 77550-636
Isopropyl myristate 98%
Isopropyl myristate

Precaución: for Research Use Only

Formula: C₁₇H₃₄O₂
MDL Number: MFCD00008982
CAS Number: 110-27-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR