Methyl (E)-4-chloro-3-methoxy-2-butenoate 95%

Supplier: Ambeed
Danger

A260180-100G A260180-1G A260180-25G A260180-5G
76801-768EA 135 USD
76801-768 76801-770 76801-772 76801-774 76801-776
Methyl (E)-4-chloro-3-methoxy-2-butenoate 95%
Methyl (E)-4-chloro-3-methoxy-2-butenoate

Precaución: for Research Use Only

Formula: C₆H₉ClO₃
MDL Number: MFCD00071562
CAS Number: 110104-60-4
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR