7-Methoxy-2-naphthol 97%

Supplier: Ambeed
Warning

Synonyms: 7-Methoxynaphthalen-2-ol

A259993-10G A259993-1G A259993-250MG A259993-5G A259993-25G
76801-760EA 89.12 USD
76801-760 76801-762 76801-764 76801-766 77668-120
7-Methoxy-2-naphthol 97%
7-Methoxy-2-naphthol

Precaución: for Research Use Only

Formula: C₁₁H₁₀O₂
MW: 174.2000 g/mol
MDL Number: MFCD00075494
CAS Number: 5060-82-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR