Ethyl-2-chlorothiazole-5-carboxylate 97%

Supplier: Ambeed
Warning

A257681-10G A257681-1G A257681-5G A257681-25G
76801-728EA 65.78 USD
76801-728 76801-730 76801-732 77601-074
Ethyl-2-chlorothiazole-5-carboxylate 97%
Ethyl-2-chlorothiazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₆H₆ClNO₂S
MDL Number: MFCD08703267
CAS Number: 81449-93-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR