Dibutyl malonate 95%
Supplier: Ambeed
|
Danger
|
Synonyms:
Malonic Acid Dibutyl Ester
A257512-1G
A257512-5G
A257512-25G
76801-724EA
7
USD
76801-724
76801-726
77668-102
Dibutyl malonate 95%
Dibutyl malonate
Precaución: for Research Use Only
Formula:
C₁₁H₂₀O₄ MW: 216.28 g/mol |
MDL Number:
MFCD00053795 CAS Number: 1190-39-2 UN: 1993 ADR: 3 Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...