Lonafarnib 99%

Supplier: Ambeed
Warning

Synonyms: SCH66336, SCH 66336, SCH-66336, (R)-4-(2-(4-(3,10-dibromo-8-chloro-6,11-dihydro-5H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-yl)piperidin-1-yl)-2-oxoethyl)piperidine-1-carboxamide, farnesyltransferase inhibitor, Sarasar, FTI inhibitor, 4-(2-(4-(8-Chloro-3,10-dibromo-6,11-dihydro-5H-benzo(5,6)cyclohepta(1,2-b)pyridin-11-yl)-1-piperidinyl)-2-oxoethyl)-1-piperidinecarboxamide

A257211-1MG A257211-5MG A257211-100MG A257211-10MG A257211-250MG
76801-718EA 21 USD
76801-718 76801-720 77668-098 77668-100 77879-479
Lonafarnib 99%
Lonafarnib

Precaución: for Research Use Only

Formula: C₂₇H₃₁Br₂ClN₄O₂
MDL Number: MFCD06795138
CAS Number: 193275-84-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR