4-(4-Morpholinylcarbonyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-(4-Morpholinylcarbonyl)benzeneboronic acid, (4-(Morpholine-4-carbonyl)phenyl)boronic acid

A256909-1G A256909-250MG A256909-5G
76801-702EA 56.23 USD
76801-702 76801-704 76801-706
4-(4-Morpholinylcarbonyl)phenylboronic acid 98%
4-(4-Morpholinylcarbonyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₄BNO₄
MW: 235.05 g/mol
MDL Number: MFCD03411952
CAS Number: 389621-84-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR