1-Hydrazinophthalazine hydrochloride 98%

Supplier: Ambeed
Danger

Synonyms: Hydralazine Hydrochloride, Hydralazine HCl, 1-Hydrazinylphthalazine hydrochloride

A256420-250MG A256420-25MG A256420-5G A256420-100MG A256420-25G A256420-1G
76801-692EA 9 USD
76801-692 76801-694 77568-482 77601-060 77668-074 77879-472
1-Hydrazinophthalazine hydrochloride 98%
1-Hydrazinophthalazine hydrochloride

Precaución: For research use only.

Formula: C₈H₈N₄
MW: 196.64 g/mol
MDL Number: MFCD00135998
CAS Number: 304-20-1
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR