Ethyl-3-chlorobenzoate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Chlorobenzoic acid ethyl ester, Ethyl 3-chlorobenzoate
A256404-100G
A256404-25G
76801-686EA
58
USD
76801-686
76801-688
76801-690
Ethyl-3-chlorobenzoate 98%
Ethyl-3-chlorobenzoate
Precaución: for Research Use Only
Formula:
C₉H₉ClO₂ |
MDL Number:
MFCD00013634 CAS Number: 1128-76-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...