5-Bromoindol-3-ylacetic acid 95%

Supplier: Ambeed
Warning

Synonyms: 5-Bromoindole-3-acetic acid, 2-(5-Bromo-1H-indol-3-yl)acetic acid

A256315-1G A256315-25G A256315-5G
76801-680EA 25.47 USD
76801-680 76801-682 76801-684
5-Bromoindol-3-ylacetic acid 95%
5-Bromoindol-3-ylacetic acid

Precaución: for Research Use Only

Formula: C₁₀H₈BrNO₂
MW: 254.08 g/mol
MDL Number: MFCD00005637
CAS Number: 40432-84-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR