Ethyl-2-(1H-indol-2-yl)acetate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-(1H-indol-2-yl)acetate

A254620-100MG A254620-1G A254620-250MG A254620-25G A254620-5G
76801-638EA 11.67 USD
76801-638 76801-640 76801-642 76801-644 76801-646
Ethyl-2-(1H-indol-2-yl)acetate 97%
Ethyl-2-(1H-indol-2-yl)acetate

Precaución: for Research Use Only

Formula: C₁₂H₁₃NO₂
MW: 203.2371 g/mol
MDL Number: MFCD08361732
CAS Number: 33588-64-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR