(R)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(2,4-dichlorophenyl)propanoic acid 96%

Supplier: Ambeed
Warning

A254175-1G A254175-250MG
76801-620EA 73.21 USD
76801-620 76801-622
(R)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(2,4-dichlorophenyl)propanoic acid 96%
(R)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(2,4-dichlorophenyl)propanoic acid

Precaución: for Research Use Only

Formula: C₂₄H₁₉Cl₂NO₄
MW: 456.32 g/mol
MDL Number: MFCD03428018
CAS Number: 511272-37-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR