(S)-N1,N3-Bis(1,3-dihydroxypropan-2-yl)-5-(2-hydroxypropanamido)-2,4,6-triiodoisophthalamide 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Iopamidol
A254011-100G
A254011-1MG
A254011-25MG
A254011-5G
76801-614EA
212.18
USD
76801-614
76801-616
76801-618
77601-040
(S)-N1,N3-Bis(1,3-dihydroxypropan-2-yl)-5-(2-hydroxypropanamido)-2,4,6-triiodoisophthalamide 98%
(S)-N1,N3-Bis(1,3-dihydroxypropan-2-yl)-5-(2-hydroxypropanamido)-2,4,6-triiodoisophthalamide
Precaución: for Research Use Only
Formula:
C₁₇H₂₂I₃N₃O₈ MW: 777.09 g/mol |
MDL Number:
MFCD00867931 CAS Number: 60166-93-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...