S,S'-Dimethyl-N-cyanodithioiminocarbonate 97%

Supplier: Ambeed
Danger

Synonyms: Dimethyl N-Cyanodithioiminocarbonate, Dimethyl N-Cyanodithiocarbonimidate

A253929-100G A253929-10G A253929-500G A253929-5G A253929-25G
76801-606EA 19 USD
76801-606 76801-608 76801-610 76801-612 77091-404
S,S'-Dimethyl-N-cyanodithioiminocarbonate 97%
S,S'-Dimethyl-N-cyanodithioiminocarbonate

Precaución: for Research Use Only

Formula: C₄H₆N₂S₂
MW: 146.23 g/mol
MDL Number: MFCD00009825
CAS Number: 10191-60-3
UN: 1759
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR