6-Bromo-4-hydroxyquinoline-3-carboxylic acid 97%

Supplier: Ambeed
Warning

A253266-10G A253266-1G A253266-250MG A253266-25G A253266-5G
76801-594EA 75 USD
76801-594 76801-596 76801-598 76801-600 76801-602
6-Bromo-4-hydroxyquinoline-3-carboxylic acid 97%
6-Bromo-4-hydroxyquinoline-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₆BrNO₃
MDL Number: MFCD01569278
CAS Number: 98948-95-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR