Fmoc-D-3-Abu-OH 98%

Supplier: Ambeed
Warning

Synonyms: (R)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)butanoic acid

A251664-10G A251664-1G A251664-25G A251664-5G
76801-576EA 155.96 USD
76801-576 76801-578 76801-580 76801-582
Fmoc-D-3-Abu-OH 98%
Fmoc-D-3-Abu-OH

Precaución: for Research Use Only

Formula: C₁₉H₁₉NO₄
MW: 325.4 g/mol
MDL Number: MFCD00270344
CAS Number: 201864-71-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR