4-Chloronaphthalen-1-amine 97%

Supplier: Ambeed
Warning

A251552-1G A251552-250MG A251552-25G A251552-5G
76801-572EA 50 USD
76801-572 76801-574 77879-460 77879-461
4-Chloronaphthalen-1-amine 97%
4-Chloronaphthalen-1-amine

Precaución: for Research Use Only

Formula: C₁₀H₈ClN
MDL Number: MFCD00004024
CAS Number: 4684-12-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR