Hexane-1,6-diyl bis(3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoate) 97%

Supplier: Ambeed
Warning

A251442-1G A251442-250MG A251442-25G A251442-5G
76801-564EA 54.11 USD
76801-564 76801-566 76801-568 76801-570
Hexane-1,6-diyl bis(3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoate) 97%
Hexane-1,6-diyl bis(3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoate)

Precaución: for Research Use Only

Formula: C₄₀H₆₂O₆
MW: 638.91 g/mol
MDL Number: MFCD00134695
CAS Number: 35074-77-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR