Malachite green chloride BS

Supplier: Ambeed
Danger

Synonyms: Malachite green, CI 42000, N,N,N′,N′-Tetramethyl-4,4′-diaminotriphenylcarbenium chloride, Basic Green 4, N-(4-((4-(Dimethylamino)phenyl)(phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-methylmethanaminium chloride, Malachite green hydrochloride

A251087-5G A251087-25G
76801-560EA 5.31 USD
76801-560 77601-028
Malachite green chloride BS
Malachite green chloride

Precaución: for Research Use Only

Formula: C₂₃H₂₅ClN₂
MW: 364.92 g/mol
MDL Number: MFCD00011751
CAS Number: 569-64-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR